missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-1-piperazineacetamide, 95%, ACROS Organics™

Brand:  Acros Organics 205610050

Code : 29

Additional Details : CAS Number : 39890-42-1 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10626432

  • £40.90 / 5g


Estimated Shipment
22-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Isopropyl-1-piperazineacetamide
39890-42-1
100.0
95%
185.27
95%
94.0
95%
C9H19N3O
MFCD00005969

Specifications

N-Isopropyl-1-piperazineacetamide
82.0° to 88.0°C
95%
94.0
Crystals and Chunks
95%
C9H19N3O
MFCD00005969
Slightly yellow
5g
39890-42-1
100.0
Authentic
95%
185.27
Glass bottle
Videos
SDS