missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Isobutyl-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60129.03

Code : NEW

Additional Details : CAS Number : 89840-80-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496598

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Isobutyl-2-nitrobenzenesulfonamide
89840-80-2
258.29

Specifications

N-Isobutyl-2-nitrobenzenesulfonamide
89840-80-2
97%
1g
Solid
258.29
Videos
SDS