missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxyphthalimide, 98%, Thermo Scientific Chemicals

Catalog Number 13458518
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
524-38-9
C8H5NO3
163.13
MFCD00005891
CFMZSMGAMPBRBE-UHFFFAOYSA-N
n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
10665
2-hydroxyisoindole-1,3-dione
ON1C(=O)C2=CC=CC=C2C1=O
This item is not returnable. View return policy
524-38-9
C8H5NO3
163.13
MFCD00005891
CFMZSMGAMPBRBE-UHFFFAOYSA-N
n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
10665
2-hydroxyisoindole-1,3-dione
ON1C(=O)C2=CC=CC=C2C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 233.0°C to 235.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Quantity 2.5 kg
Beilstein ,5
Fieser 01,485; 07,177
Merck Index 15,4876
Solubility Information Solubility in water: slightly soluble
Formula Weight 163.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Hydroxyphthalimide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-Hydroxyphthalimide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'