missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog Number p-146291
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
118-29-6
C9H7NO3
177.16
MFCD00005899
MNSGOOCAMMSKGI-UHFFFAOYSA-N
n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
8354
CHEBI:38816
2-(hydroxymethyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CO
This item is not returnable. View return policy
118-29-6
C9H7NO3
177.16
MFCD00005899
MNSGOOCAMMSKGI-UHFFFAOYSA-N
n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
8354
CHEBI:38816
2-(hydroxymethyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 140°C to 145°C
Color White
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 475
Fieser 01,484; 08,253
Formula Weight 177.16
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-(Hydroxymethyl)phthalimide, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'