missing translation for 'onlineSavingsMsg'
Learn More

N'-Hydroxycyclopropanecarboximidamide, 97%, Thermo Scientific™

Catalog Number p-3076407
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
51285-13-3
C4H8N2O
100.121
MFCD07772876
OMCUPXRCMTUDHI-UHFFFAOYSA-N
n-hydroxycyclopropanecarboximidamide, z-n'-hydroxycyclopropanecarboxamidine, n-hydroxycyclopropanecarboxamidine, n'-hydroxycyclopropanecarboxamidine, z-n'-hydroxycyclopropanec arboximidamide, cyclopropanecarboxamidoxime, n-hydroxy-cyclopropanecarboximidamide, n'-hydroxycyclopropane-carboximidamide, z-n'-hydroxycyclopropanecarboximidamide, n'-hydroxycyclopropanecarboximideamide
9582826
N'-hydroxycyclopropanecarboximidamide
C1CC1C(=NO)N
This item is not returnable. View return policy
51285-13-3
C4H8N2O
100.121
MFCD07772876
OMCUPXRCMTUDHI-UHFFFAOYSA-N
n-hydroxycyclopropanecarboximidamide, z-n'-hydroxycyclopropanecarboxamidine, n-hydroxycyclopropanecarboxamidine, n'-hydroxycyclopropanecarboxamidine, z-n'-hydroxycyclopropanec arboximidamide, cyclopropanecarboxamidoxime, n-hydroxy-cyclopropanecarboximidamide, n'-hydroxycyclopropane-carboximidamide, z-n'-hydroxycyclopropanecarboximidamide, n'-hydroxycyclopropanecarboximideamide
9582826
N'-hydroxycyclopropanecarboximidamide
C1CC1C(=NO)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material N';-Hydroxycyclopropanecarboximidamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'