Learn More
N'-Hydroxycyclopropanecarboximidamide, 97%, Maybridge
Brand: Maybridge MO01104EA
Code : 31
Additional Details : CAS Number : 51285-13-3 Weight : 0.06000kg Transport : UN number : 2810 Chem class : 1 Pack group : III
Quantity | 10g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
N';-Hydroxycyclopropanecarboximidamide | |
97.0 | |
97% | |
100.12 | |
N«-Hydroxycyclopropanecarboximidamide (IUPAC) |
51285-13-3 | |
97% | |
C4H8N2O | |
MFCD07772876 |
Specifications
N';-Hydroxycyclopropanecarboximidamide | |
51285-13-3 | |
Amber Glass Bottle | |
97% | |
100.12 | |
N«-Hydroxycyclopropanecarboximidamide (IUPAC) |
10g | |
97.0 | |
97% | |
C4H8N2O | |
MFCD07772876 |