missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Catalog Number 10457432
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
613-92-3
C7H8N2O
136.15
MFCD00031485 MFCD00474011
MXOQNVMDKHLYCZ-UHFFFAOYSA-N
benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
7259353
CHEBI:83354
N\C(=N/O)C1=CC=CC=C1
This item is not returnable. View return policy
613-92-3
C7H8N2O
136.15
MFCD00031485 MFCD00474011
MXOQNVMDKHLYCZ-UHFFFAOYSA-N
benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
7259353
CHEBI:83354
N\C(=N/O)C1=CC=CC=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5C(NH2)=NOH
Quantity 10 g
IUPAC Name N'-hydroxybenzenecarboximidamide
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material N'-Hydroxybenzenecarboximidamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'