missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-2,2-dimethylpropanimidamide, 95%, Thermo Scientific Chemicals

Catalog Number 11925671
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
42956-75-2
C5H12N2O
116.16
CVOGFMYWFRFWEQ-UHFFFAOYSA-N
n-hydroxypivalimidamide, 1z-n'-hydroxy-2,2-dimethylpropanimidamide, n-hydroxy-2,2-dimethylpropanimidamide, n-hydroxypivalamidine, z-n'-hydroxy-2,2-dimethylpropanimidamide, hydroxydimethylpropanimidamide, z-n'-hydroxypivalimidamide, z-n'-hydroxy-2,2-dimethylpropimidamide
9582836
N'-hydroxy-2,2-dimethylpropanimidamide
CC(C)(C)C(=NO)N
This item is not returnable. View return policy
42956-75-2
C5H12N2O
116.16
CVOGFMYWFRFWEQ-UHFFFAOYSA-N
n-hydroxypivalimidamide, 1z-n'-hydroxy-2,2-dimethylpropanimidamide, n-hydroxy-2,2-dimethylpropanimidamide, n-hydroxypivalamidine, z-n'-hydroxy-2,2-dimethylpropanimidamide, hydroxydimethylpropanimidamide, z-n'-hydroxypivalimidamide, z-n'-hydroxy-2,2-dimethylpropimidamide
9582836
N'-hydroxy-2,2-dimethylpropanimidamide
CC(C)(C)C(=NO)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 108.6°C to 113.7°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 116.16
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Hydroxy-2, 2-dimethylpropanimidamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'