missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl formate, 97%, Thermo Scientific™

Catalog Number p-7058307
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
629-33-4
C7H14O2
130.187
MFCD00046146
OUGPMNMLWKSBRI-UHFFFAOYSA-N
n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
61177
hexyl formate
CCCCCCOC=O
This item is not returnable. View return policy
629-33-4
C7H14O2
130.187
MFCD00046146
OUGPMNMLWKSBRI-UHFFFAOYSA-N
n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
61177
hexyl formate
CCCCCCOC=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point -63°C
Boiling Point 155°C
Flash Point 37°C (98°F)
Quantity 50 g
UN Number UN3272
Beilstein 1701629
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material n-Hexyl formate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'