missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
200 mL
800 mL
Description
n-Hexane is used in the formulation of glues for shoes, leather products, and roofing, as a cleaning agent, de-greasing agent. It is used to extract edible oils from seeds and vegetables, a non-polar solvent in chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Melting Point | -95°C |
Density | 0.659 g/mL |
Boiling Point | 69°C |
Quantity | 200 mL |
Flash Point | -23°C (-9°F) |
Odor | Weak, characteristic |
Packaging | ChemSeal™ Bottle |
Physical Form | Liquid |
Refractive Index | 1.375 |
UN Number | UN1208 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'