missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals

Catalog Number 15401787
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC

For Pesticide Residue, Environmental and GC Analysis

Pesticide residue, environmental and GC analysis.n-Hexane is used in spectrophotometry and environmental testing. It is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 95°C
Color Colorless
Appearance Colorless liquid
Density 0.659
Boiling Point 69°C
Quantity 4 x 1 L
Flash Point -23°C (-9F)
Ignition Point 240°C (464°F)
Odor Weak, characteristic
Physical Form Liquid
Refractive Index 1.375
Assay Percent Range >95%
Beilstein 1730733
Solubility Information Very soluble in ethanol,ethyl ether and chloroform.
Formula Weight 86.18
Grade Environmental
Chemical Name or Material n-Hexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'