missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane-d{14}, 99%(Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051950
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
21666-38-6
C6H14
100.26
MFCD00037562
VLKZOEOYAKHREP-ZLKPZJALSA-N
hexane-d14, tetradecadeuterohexane, n-hexane-d 14, 2h14 hexane, 2 h?? hexane, hexane-d14, 99 atom % d
140854
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecadeuteriohexane
[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
21666-38-6
C6H14
100.26
MFCD00037562
VLKZOEOYAKHREP-ZLKPZJALSA-N
hexane-d14, tetradecadeuterohexane, n-hexane-d 14, 2h14 hexane, 2 h?? hexane, hexane-d14, 99 atom % d
140854
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecadeuteriohexane
[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

n-Hexane-d{14} is a deuterium labeled n-hexane which is used as a organic intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 0.767 g/mL
Flash Point −23°C (−9°F)
Odor Characteristic
Quantity 1 g
Melting Point -95°C
Boiling Point 69°C
UN Number UN1208
Solubility Information Mixes with most organic solvents,chloroform,ether and alcohol. Insoluble in water.
Formula Weight 100.29
Percent Purity 99%
Chemical Name or Material n-Hexane-d{14}, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'