missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals

Catalog Number p-3758160
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
540-10-3
C32H64O2
480.86
MFCD00053739
PXDJXZJSCPSGGI-UHFFFAOYSA-N
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
10889
CHEBI:75584
hexadecyl hexadecanoate
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
This item is not returnable. View return policy
540-10-3
C32H64O2
480.86
MFCD00053739
PXDJXZJSCPSGGI-UHFFFAOYSA-N
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
10889
CHEBI:75584
hexadecyl hexadecanoate
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 52°C to 56°C
Color White
Boiling Point 360°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)14COO(CH2)15CH3
Quantity 5 g
Merck Index 15, 2030
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 480.86
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material n-Hexadecyl palmitate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'