missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl mercaptan, 90%, technical, Thermo Scientific Chemicals

Catalog Number 10645542
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2917-26-2
C16H34S
258.5
MFCD00011677
ORTRWBYBJVGVQC-UHFFFAOYSA-N
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan
18015
hexadecane-1-thiol
CCCCCCCCCCCCCCCCS
This item is not returnable. View return policy
2917-26-2
C16H34S
258.5
MFCD00011677
ORTRWBYBJVGVQC-UHFFFAOYSA-N
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan
18015
hexadecane-1-thiol
CCCCCCCCCCCCCCCCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 18°C to 20°C
Color Colorless or White to Yellow
Density 0.8400g/mL
Boiling Point 184°C to 191°C (7.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)15SH
Refractive Index 1.4610 to 1.463
Quantity 100 mL
Beilstein 01, 430
Specific Gravity 0.84
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethyl alcohol,carbon tetrachl,soluble in ether
Formula Weight 258.5
Percent Purity 90%
Grade Technical
Physical Form Liquid or Waxy Solid
Chemical Name or Material n-Hexadecyl mercaptan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'