missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, 99%, pure, ACROS Organics™

Brand:  Acros Organics 120460100

Code : NEW

Additional Details : CAS Number : 544-76-3 Weight : 7.73000kg

Quantity 10L

 View more versions of this product

Product Code. 15234614

  • £1087.10 / 10 L


EU Stock 5
Add to basket

Specifications

Chemical Identifiers

n-Hexadecane
98.5
Pure
99%
CH3(CH2)14CH3
MFCD00008998
544-76-3
100.0
99%
C16H34
226.44

Specifications

n-Hexadecane
544-76-3
100.0
Pure
99%
C16H34
CH3(CH2)14CH3
MFCD00008998
0.773
287.0°C
Clear colorless
10L
98.5
Authentic
Liquid
99%
Glass Bottle
226.44
1.4330 to 1.4350
0.7730g/mL
18.0°C
135°C
Videos