missing translation for 'onlineSavingsMsg'
Learn More

n-Heptyltriphenylphosphonium bromide, 99%, ACROS Organics™

Brand:  Acros Organics 259260500

Code : 29

Additional Details : CAS Number : 13423-48-8 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10482431

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

n-Heptyltriphenylphosphonium bromide
13423-48-8
100.0
99%
CH3(CH2)6P(C6H5)3Br
MFCD00050249
99%
98.5
99%
C25H30BrP
441.38

Specifications

n-Heptyltriphenylphosphonium bromide
99%
98.5
Authentic
99%
C25H30BrP
CH3(CH2)6P(C6H5)3Br
MFCD00050249
White
50g
13423-48-8
100.0
Fine Crystalline Powder
99%
Glass bottle
441.38
175.0° to 178.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Heptyltriphenylphosphonium bromide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS