missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals

Catalog Number p-216851
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
TRUSTED_SUSTAINABILITY
Melting Point -91.0°C
CAS Min % 99.4
Color Colorless to Yellow
CAS Max % 100.0
Density 0.6830g/mL
Boiling Point 98.0°C
Quantity 1 L
Flash Point −4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3850 to 1.3890
Assay Percent Range 99+%
Residue after Evaporation 0.0003 % max.
Specific Gravity 0.683
Linear Formula CH3(CH2)5CH3
Viscosity 0.4 mPa.s (20°C)
Beilstein 01, 154
Water 0.01% Max. (Coulometric)
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Percent Purity ≥99%
Grade Residue Analysis
Chemical Name or Material n-Heptane, ECD Tested, For Pesticide Analysis, Sulfur Low
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'