missing translation for 'onlineSavingsMsg'
Learn More

n-Heneicosane, 98%, Thermo Scientific Chemicals

Catalog Number 10602952
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-94-7
C21H44
296.58
MFCD00009346
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
12403
CHEBI:32931
henicosane
CCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-94-7
C21H44
296.58
MFCD00009346
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
12403
CHEBI:32931
henicosane
CCCCCCCCCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 39°C to 43°C
Color White
Boiling Point 100°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)19CH3
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether
Formula Weight 296.58
Percent Purity 98%
Physical Form Crystalline Chunks, Crystals, or Flakes
Chemical Name or Material n-Heneicosane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'