missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-S-benzyl-L-cysteine, 98%, Thermo Scientific™

Catalog Number 15409468
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
53298-33-2
C25H23NO4S
433.522
MFCD00065633
AKXYVGAAQGLAMD-QHCPKHFHSA-N
fmoc-cys bzl-oh, fmoc-s-benzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzylthio propanoic acid, l-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-phenylmethyl, 2r-3-benzylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-s-benzyl-l-cys, pubchem12391, fmoc-l-cys bzl-oh, ksc925c9h, n-fmoc-s-phenylmethyl-l-cysteine
11750859
(2R)-3-benzylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
This item is not returnable. View return policy
53298-33-2
C25H23NO4S
433.522
MFCD00065633
AKXYVGAAQGLAMD-QHCPKHFHSA-N
fmoc-cys bzl-oh, fmoc-s-benzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzylthio propanoic acid, l-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-phenylmethyl, 2r-3-benzylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-s-benzyl-l-cys, pubchem12391, fmoc-l-cys bzl-oh, ksc925c9h, n-fmoc-s-phenylmethyl-l-cysteine
11750859
(2R)-3-benzylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 433.52
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Fmoc-S-benzyl-L-cysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'