missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-S-(4-methylbenzyl)-L-cysteine, 95%

Brand:  Alfa Aesar™ H63950.06

Code : NEW

Additional Details : CAS Number : 136050-67-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15420949

  • £76.96 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-S-(4-methylbenzyl)-L-cysteine
447.55
6670642
136050-67-4
MFCD00065634
Fmoc-cys(4-mBzl)-OH

Specifications

N-Fmoc-S-(4-methylbenzyl)-L-cysteine
Solid
447.55
6670642
150° to 158°C
136050-67-4
95%
MFCD00065634
Fmoc-cys(4-mBzl)-OH
5g
Videos
SDS