missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-N-methyl-L-threonine, 99%, Thermo Scientific Chemicals

Catalog Number 15429718
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
117106-20-4
C24H29NO5
411.50
MFCD02094431
VIUVLZHFMIFLHU-VFNWGFHPSA-N
fmoc-n-me-thr tbu-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy butanoic acid, n-fmoc-n-methyl-o-tert-butyl-l-threonine, fmoc-n-methyl-o-t-butyl-l-threonine, pubchem19052, fmoc-n-me-thr but-oh, fmoc-nalpha-methyl-o-t-butyl-l-threonine, fmoc-n-alpha-methyl-o-t-butyl-l-threonine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-threonine
7010372
(2S,3R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]butanoic acid
C[C@@H](OC(C)(C)C)[C@H](N(C)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
This item is not returnable. View return policy
117106-20-4
C24H29NO5
411.50
MFCD02094431
VIUVLZHFMIFLHU-VFNWGFHPSA-N
fmoc-n-me-thr tbu-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy butanoic acid, n-fmoc-n-methyl-o-tert-butyl-l-threonine, fmoc-n-methyl-o-t-butyl-l-threonine, pubchem19052, fmoc-n-me-thr but-oh, fmoc-nalpha-methyl-o-t-butyl-l-threonine, fmoc-n-alpha-methyl-o-t-butyl-l-threonine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-threonine
7010372
(2S,3R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]butanoic acid
C[C@@H](OC(C)(C)C)[C@H](N(C)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 411.49
Percent Purity 99%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Fmoc-O-tert-butyl-N-methyl-L-threonine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'