missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-D-allo-threonine, 98%, Thermo Scientific Chemicals

Catalog Number p-7057125
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
170643-02-4
C23H27NO5
397.47
MFCD00077074
LZOLWEQBVPVDPR-JLTOFOAXSA-N
fmoc-d-allo-thr tbu-oh, 2r,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-d-allo-threonine tbu-oh, nalpha-9-fluorenylmethoxycarbonyl-o-t-butyl-d-allo-threonine, n-fmoc-o-tert-butyl-d-allothreonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-d-allothreonine, o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-d-allothreonine, o-tert-butyl-n-9h-fluoren-9-yl methoxy carbonyl-d-allothreonine, 2r,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, d-allothreonine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl
10949330
(2R,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
C[C@@H](OC(C)(C)C)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
This item is not returnable. View return policy
170643-02-4
C23H27NO5
397.47
MFCD00077074
LZOLWEQBVPVDPR-JLTOFOAXSA-N
fmoc-d-allo-thr tbu-oh, 2r,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-d-allo-threonine tbu-oh, nalpha-9-fluorenylmethoxycarbonyl-o-t-butyl-d-allo-threonine, n-fmoc-o-tert-butyl-d-allothreonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-d-allothreonine, o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-d-allothreonine, o-tert-butyl-n-9h-fluoren-9-yl methoxy carbonyl-d-allothreonine, 2r,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, d-allothreonine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl
10949330
(2R,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
C[C@@H](OC(C)(C)C)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 397.47
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Fmoc-O-tert-butyl-D-allo-threonine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'