missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-benzyl-D-tyrosine, 95%, Thermo Scientific™

Catalog Number 15432789
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
138775-48-1
C31H27NO5
493.56
MFCD00235837
REHSJSKPWIOKIJ-KSTXGFNDNA-N
fmoc-d-tyr bzl-oh, fmoc-o-benzyl-d-tyr, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, 2r-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-o-benzyl-l-tyr, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-d-tyrosine, o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-d-tyrosine, d-tyrosine,n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl
40428316
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
OC(=O)[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
This item is not returnable. View return policy
138775-48-1
C31H27NO5
493.56
MFCD00235837
REHSJSKPWIOKIJ-KSTXGFNDNA-N
fmoc-d-tyr bzl-oh, fmoc-o-benzyl-d-tyr, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, 2r-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-o-benzyl-l-tyr, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-d-tyrosine, o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-d-tyrosine, d-tyrosine,n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl
40428316
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
OC(=O)[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 439.56
Percent Purity 95%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Fmoc-O-benzyl-D-tyrosine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'