missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-α-FMOC-N-Trityl-L-histidine, 98%

Catalog Number 10511862
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
109425-51-6
C40H33N3O4
619.72
XXMYDXUIZKNHDT-QNGWXLTQSA-N
fmoc-his trt-oh, n-fmoc-n'-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, a-fmoc-n im-trityl-l-histidine, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, ambotzfaa1090
11422193
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
This item is not returnable. View return policy
109425-51-6
C40H33N3O4
619.72
XXMYDXUIZKNHDT-QNGWXLTQSA-N
fmoc-his trt-oh, n-fmoc-n'-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, a-fmoc-n im-trityl-l-histidine, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, ambotzfaa1090
11422193
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 133.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation 83.3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Residual Solvents 5% max. 2-propanol, THF
Formula Weight 619.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-FMOC-N-Trityl-L-histidine
Show More Show Less
compliance-icons
Product Identifier
  • N-¢(9H-Fluoren-6-ylmethoxy)carbonyl!-3-(triphenylmethyl)-L-histidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'