missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-N-methylglycine, 98%, Thermo Scientific Chemicals
36.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in DMF.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in DMF.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications
Quantity | 5 g |
Beilstein | 6660958 |
Solubility Information | Soluble in DMF. |
Formula Weight | 311.33 |
Percent Purity | 98% |
Chemical Name or Material | N-Fmoc-N-methylglycine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'