missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-L-threonine, 95%

Brand:  Alfa Aesar™ H63376.03

Code : NEW

Additional Details : CAS Number : 252049-06-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15440039

  • £32.33 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-N-methyl-L-threonine
355.38
Fmoc-N-Me-Thr-OH
252049-06-2
MFCD02094575

Specifications

N-Fmoc-N-methyl-L-threonine
Solid
355.38
Fmoc-N-Me-Thr-OH
252049-06-2
95%
MFCD02094575
1g
Videos
SDS