missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-L-threonine, 95%

Brand:  Alfa Aesar™ H63376.06

Code : NEW

Additional Details : CAS Number : 252049-06-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450039

  • £161.12 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-N-methyl-L-threonine
355.38
Fmoc-N-Me-Thr-OH
252049-06-2
MFCD02094575

Specifications

N-Fmoc-N-methyl-L-threonine
Solid
355.38
Fmoc-N-Me-Thr-OH
252049-06-2
95%
MFCD02094575
5g
Videos
SDS