missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-L-glutamic acid 5-tert-butyl ester, 98%

Brand:  Alfa Aesar™ H63814.03

Code : NEW

Additional Details : CAS Number : 200616-40-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438536

  • £146.28 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-N-methyl-L-glutamic acid 5-tert-butyl ester
439.5
Fmoc-N-Me-Glu(OtBu)-OH
200616-40-6
MFCD00237028

Specifications

N-Fmoc-N-methyl-L-glutamic acid 5-tert-butyl ester
Solid
439.5
Fmoc-N-Me-Glu(OtBu)-OH
200616-40-6
98%
MFCD00237028
1g
Videos
SDS