missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-D-leucine, 97%

Brand:  Alfa Aesar™ H63895.06

Code : NEW

Additional Details : CAS Number : 103478-63-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410859

  • £141.30 / 5g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Description

Description

Applications
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Chemical Identifiers

N-Fmoc-N-methyl-D-leucine
367.44
7350417
103478-63-3
MFCD00235877
Fmoc-N-Me-D-Leu-OH

Specifications

N-Fmoc-N-methyl-D-leucine
Powder
367.44
7350417
Off-white
103478-63-3
97%
MFCD00235877
Fmoc-N-Me-D-Leu-OH
5g
Videos
SDS