missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-propargylglycine, 95%, Thermo Scientific Chemicals

Catalog Number p-7034485
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
198561-07-8
C20H17NO4
335.359
MFCD01075095
DJGMNCKHNMRKFM-SFHVURJKSA-N
fmoc-l-propargylglycine, fmoc-pra-oh, fmoc-propargyl-gly-oh, s-2-fmoc-amino-4-pentynoic acid, s-n-fmoc-2-2'-propynyl glycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-ynoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-ynoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, 4-pentynoicacid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
2734461
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-ynoic acid
C#CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
198561-07-8
C20H17NO4
335.359
MFCD01075095
DJGMNCKHNMRKFM-SFHVURJKSA-N
fmoc-l-propargylglycine, fmoc-pra-oh, fmoc-propargyl-gly-oh, s-2-fmoc-amino-4-pentynoic acid, s-n-fmoc-2-2'-propynyl glycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-ynoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-ynoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, 4-pentynoicacid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
2734461
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-ynoic acid
C#CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Fmoc-propargyl-Gly-OH is an Fmoc protected glycine derivative useful for solid phase peptide synthesis techniques. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. N-Fmoc-L-propargylglycine is a non-natural amino acids used for the triazole cross-link.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-propargyl-Gly-OH is an Fmoc protected glycine derivative useful for solid phase peptide synthesis techniques. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. N-Fmoc-L-propargylglycine is a non-natural amino acids used for the triazole cross-link.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 9151699
Formula Weight 335.36
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-propargylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'