missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-L-proline pentafluorophenyl ester, 98%

Brand:  Alfa Aesar™ H63457.03

Code : NEW

Additional Details : CAS Number : 86060-90-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15480159

  • £14.80 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-L-proline pentafluorophenyl ester
503.42
4731096
86060-90-4
MFCD00065672
Fmoc-Pro-OPfp

Specifications

N-Fmoc-L-proline pentafluorophenyl ester
Solid
503.42
4731096
127° to 129°C
86060-90-4
98%
MFCD00065672
Fmoc-Pro-OPfp
1g
Videos
SDS