missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-L-isoleucine pentafluorophenyl ester, 98%

Brand:  Alfa Aesar™ H63691.03

Code : NEW

Additional Details : CAS Number : 86060-89-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430519

  • £15.16 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-L-isoleucine pentafluorophenyl ester
519.46
4221390
86060-89-1
MFCD00077063
Fmoc-Ile-OPfp

Specifications

N-Fmoc-L-isoleucine pentafluorophenyl ester
Solid
519.46
4221390
1g
86060-89-1
98%
MFCD00077063
Fmoc-Ile-OPfp
Videos
SDS