missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-homoarginine, 96%, Thermo Scientific™

Catalog Number p-7054810
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
776277-76-0
C22H26N4O4
410.474
MFCD01861360
QRELIJBJYZHTQU-IBGZPJMESA-N
fmoc-hoarg-oh, fmoc-homoarg-oh, fmoc-har-oh, fmoc-l-homoarginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-guanidinohexanoic acid, fmoc-homoarg, 2s-6-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n∼6∼-diaminomethylidene-n∼2∼-9h-fluoren-9-yl methoxy carbonyl-l-lysine, 2s-6-diaminomethylidene amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
40418724
(2S)-6-(diaminomethylideneamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=C(N)N)C(=O)O
This item is not returnable. View return policy
776277-76-0
C22H26N4O4
410.474
MFCD01861360
QRELIJBJYZHTQU-IBGZPJMESA-N
fmoc-hoarg-oh, fmoc-homoarg-oh, fmoc-har-oh, fmoc-l-homoarginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-guanidinohexanoic acid, fmoc-homoarg, 2s-6-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n∼6∼-diaminomethylidene-n∼2∼-9h-fluoren-9-yl methoxy carbonyl-l-lysine, 2s-6-diaminomethylidene amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
40418724
(2S)-6-(diaminomethylideneamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=C(N)N)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 410.47
Percent Purity 96%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Fmoc-L-homoarginine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'