missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-methyl ester, 97%, Thermo Scientific Chemicals

Catalog Number 15400019
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
145038-50-2
C21H21NO6
383.40
MFCD01076132
RULINAWEYRMHHQ-UHFFFAOYNA-N
fmoc-glu ome-oh, fmoc-l-glutamic acid gamma-methyl ester, fmoc-l-glu me-oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methoxy-5-oxopentanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-fmoc-l-glutamic acid alpha-methyl ester, ambotzfaa1720, pubchem13156, fmoc-l-glutamic acid, a-methyl ester, n-fmoc-l-glutamic acid 5-methyl ester
51340556
COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
145038-50-2
C21H21NO6
383.40
MFCD01076132
RULINAWEYRMHHQ-UHFFFAOYNA-N
fmoc-glu ome-oh, fmoc-l-glutamic acid gamma-methyl ester, fmoc-l-glu me-oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methoxy-5-oxopentanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-fmoc-l-glutamic acid alpha-methyl ester, ambotzfaa1720, pubchem13156, fmoc-l-glutamic acid, a-methyl ester, n-fmoc-l-glutamic acid 5-methyl ester
51340556
COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methoxy-5-oxopentanoic acid
Formula Weight 383.39
Percent Purity 97%
Quantity 5 g
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 5-methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'