missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homovaline, 95%, Thermo Scientific Chemicals

Catalog Number p-7034496
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
172695-33-9
C21H23NO4
353.42
MFCD01862853
KFDMTAMWOIQTOB-LJQANCHMSA-N
fmoc-l-beta-homovaline, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-leu-oh, fmoc-l-, a-leucine, fmoc-l-beta-leucine, fmoc-, a-hoval-oh
2761560
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
172695-33-9
C21H23NO4
353.42
MFCD01862853
KFDMTAMWOIQTOB-LJQANCHMSA-N
fmoc-l-beta-homovaline, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-leu-oh, fmoc-l-, a-leucine, fmoc-l-beta-leucine, fmoc-, a-hoval-oh
2761560
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 7388393
Solubility Information Slightly soluble in water.
Formula Weight 353.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homovaline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'