missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-D-tyrosine, 97%

Brand:  Alfa Aesar™ H66740.MD

Code : NEW

Additional Details : CAS Number : 112883-29-1 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15412819

  • £13.70 / 250mg


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-D-tyrosine
403.43
Fmoc-D-Tyr-OH
112883-29-1
MFCD00171384

Specifications

N-Fmoc-D-tyrosine
Solid
403.43
Fmoc-D-Tyr-OH
175° to 180°C
112883-29-1
97%
MFCD00171384
White
250mg
Videos
SDS