missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-D-alanine pentafluorophenyl ester, 98%

Brand:  Alfa Aesar™ H63713.06

Code : NEW

Additional Details : CAS Number : 125043-04-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450549

  • £147.34 / 5g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-D-alanine pentafluorophenyl ester
477.38
Fmoc-D-Ala-OPfp
125043-04-1
MFCD00153372

Specifications

N-Fmoc-D-alanine pentafluorophenyl ester
Solid
477.38
Fmoc-D-Ala-OPfp
125043-04-1
98%
MFCD00153372
5g
Videos
SDS