missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-4-nitro-D-phenylalanine, 95%

Brand:  Alfa Aesar™ H63646.03

Code : NEW

Additional Details : CAS Number : 177966-63-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410449

  • £19.93 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Fmoc-4-nitro-D-phenylalanine
432.43
8873625
177966-63-1
MFCD00237029
Fmoc-D-Phe(4-NO2)-OH

Specifications

N-Fmoc-4-nitro-D-phenylalanine
Solid
432.43
8873625
1g
177966-63-1
95%
MFCD00237029
Fmoc-D-Phe(4-NO2)-OH
Videos
SDS