missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-cyclohexyl-L-alanine, 98%, Thermo Scientific Chemicals

Catalog Number 15460869
33.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
135673-97-1
C24H27NO4
393.48
MFCD00065614
HIJAUEZBPWTKIV-UHFFFAOYNA-N
fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine
978326
(2S)-3-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
135673-97-1
C24H27NO4
393.48
MFCD00065614
HIJAUEZBPWTKIV-UHFFFAOYNA-N
fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine
978326
(2S)-3-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 7052264
Formula Weight 393.48
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Fmoc-3-cyclohexyl-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'