missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-2-octyl-L-glycine, 98%, Thermo Scientific Chemicals

Catalog Number 15442689
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
193885-59-5
C25H31NO4
409.526
MFCD01311753
LSMLSLHVRMSYPT-QHCPKHFHSA-N
fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
23257324
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
193885-59-5
C25H31NO4
409.526
MFCD01311753
LSMLSLHVRMSYPT-QHCPKHFHSA-N
fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
23257324
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

A homolog of lipophilic building block for SAR studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A homolog of lipophilic building block for SAR studies.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 140°C to 143°C
Color White
Solubility Information Slightly soluble in water.
Formula Weight 409.53
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Fmoc-2-octyl-L-glycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'