Learn More
N-Ethylmaleimide, 99+%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+ dependent internucleosomal DNA fragmentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 43°C to 46°C |
Boiling Point | 209°C to 211°C |
Flash Point | 73°C (163°F) |
Assay Percent Range | >99+% |
Quantity | 1 g |
Beilstein | 112448 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in ethanol,water,chloroform,methanol and toluene. |
Formula Weight | 125.13 |
Physical Form | Crystalline |
Show More |
RUO – Research Use Only