missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Ethyl-o-toluidine, 98%

Brand:  Alfa Aesar™ A11315.AE

Code : NEW

Additional Details : CAS Number : 94-68-8 Weight : 0.01000kg Transport : UN number : 2754 Chem class : 1 Pack group : II

Quantity 100mL

Product Code. 11403714

  • £19.80 / 100mL


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Specifications

N-Ethyl-o-toluidine
98%
135.21
UN2754
2689722
98°C (208°F)
94-68-8
C9H13N
MFCD00026695
218°C
100mL
2-Ethylaminotoluene
Videos
SDS