missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-Ethyl-N-methylbenzylamine, 99%
Click to view available options
Quantity:
10 g
50 g
Specifications
Density | 1.03 |
Boiling Point | 196°C |
Refractive Index | 1.4995 |
Quantity | 10 g |
UN Number | UN2735 |
Beilstein | 2082378 |
Formula Weight | 149.24 |
Percent Purity | 99% |
Chemical Name or Material | N-Ethyl-N-methylbenzylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'