missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Ethyl-3-methyl-N-(3-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59043.03

Code : NEW

Additional Details : CAS Number : 958996-84-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495218

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Ethyl-3-methyl-N-(3-methylphenyl)benzamide
958996-84-4
253.34
MFCD22683039

Specifications

N-Ethyl-3-methyl-N-(3-methylphenyl)benzamide
Liquid
253.34
Pale yellow
958996-84-4
97%
MFCD22683039
1g
Videos
SDS