missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech., Thermo Scientific Chemicals

Catalog Number 10512354
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7570-45-8
C15H13NO
223.28
MFCD00004963
QGJXVBICNCIWEL-UHFFFAOYSA-N
9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
82055
9-ethylcarbazole-3-carbaldehyde
CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2
This item is not returnable. View return policy
7570-45-8
C15H13NO
223.28
MFCD00004963
QGJXVBICNCIWEL-UHFFFAOYSA-N
9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
82055
9-ethylcarbazole-3-carbaldehyde
CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 92.0
CAS Max % 100.0
Melting Point 85°C to 91°C
Color Yellow to Ochre
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Quantity 500 g
Formula Weight 223.27
Percent Purity 94%
Grade Technical
Physical Form Crystalline Powder or Granules
Chemical Name or Material N-Ethyl-3-carbazolecarboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'