missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech., ACROS Organics™

£76.05 - £777.00

Chemical Identifiers

CAS 7570-45-8
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.27
MDL Number MFCD00004963
InChI Key QGJXVBICNCIWEL-UHFFFAOYSA-N
Synonym 9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
PubChem CID 82055
IUPAC Name 9-ethylcarbazole-3-carbaldehyde
SMILES CCN1C2=C(C=C(C=C2)C=O)C3=CC=CC=C31
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10685792
SDS View Documents
Acros Organics
166030250
25g Glass bottle
£76.05
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 2 Please to purchase this item. Need a web account? Register with us today!
10071920
SDS View Documents
Acros Organics
166031000
100g Glass bottle
£222.00
100g
View alternative products
Save Up to 
»
Estimated Shipment
16-12-2020
Please to purchase this item. Need a web account? Register with us today!
10512354
SDS View Documents
Acros Organics
166035000
500g Glass bottle
£777.00
500g
View alternative products
Save Up to 
»
Estimated Shipment
16-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7570-45-8
223.27
QGJXVBICNCIWEL-UHFFFAOYSA-N
82055
CCN1C2=C(C=C(C=C2)C=O)C3=CC=CC=C31
C15H13NO
MFCD00004963
9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
9-ethylcarbazole-3-carbaldehyde

Specifications

N-Ethyl-3-carbazolecarboxaldehyde
Glass bottle
85°C to 91°C
7570-45-8
100.0
C15H13NO
9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
CCN1C2=C(C=C(C=C2)C=O)C3=CC=CC=C31
223.27
223.27
94%
Authentic
Yellow to Ochre
25g
92.0
92% min. (HPLC)
MFCD00004963
QGJXVBICNCIWEL-UHFFFAOYSA-N
9-ethylcarbazole-3-carbaldehyde
82055
Crystalline Powder or Granules
Technical
SDS