missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Ethyl-2-fluoro-N-(3-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59204.03

Code : NEW

Additional Details : CAS Number : 881596-57-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495438

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Ethyl-2-fluoro-N-(3-methylphenyl)benzamide
881596-57-2
257.31
MFCD07184764

Specifications

N-Ethyl-2-fluoro-N-(3-methylphenyl)benzamide
Liquid
257.31
Orange
881596-57-2
97%
MFCD07184764
1g
Videos
SDS