missing translation for 'onlineSavingsMsg'
Learn More

n-Decanophenone, 99%, ACROS Organics™

Catalog Number p-3778110
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6048-82-4
C16H24O
232.37
MFCD00009582
QQXJNLYVPPBERR-UHFFFAOYSA-N
decanophenone, 1-decanone, 1-phenyl, n-decanophenone, capriphenone, ketone, nonyl phenyl, ketone, nonenyl phenyl, nonyl phenyl ketone, 1-phenyl-1-decanone, acmc-209mkb
80148
1-phenyldecan-1-one
CCCCCCCCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
6048-82-4
C16H24O
232.37
MFCD00009582
QQXJNLYVPPBERR-UHFFFAOYSA-N
decanophenone, 1-decanone, 1-phenyl, n-decanophenone, capriphenone, ketone, nonyl phenyl, ketone, nonenyl phenyl, nonyl phenyl ketone, 1-phenyl-1-decanone, acmc-209mkb
80148
1-phenyldecan-1-one
CCCCCCCCCC(=O)C1=CC=CC=C1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 232.37
Percent Purity ≥98.5%
Physical Form Crystalline Solid
Chemical Name or Material n-Decanophenone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'