missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclopropyl-4-fluorobenzylamine, 97%, Thermo Scientific™

Catalog Number p-7037464
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
625437-46-9
C10H12FN
165.21
MFCD07330007
YKNFXOCMGURJMK-UHFFFAOYSA-N
n-4-fluorobenzyl cyclopropanamine, cyclopropyl-4-fluoro-benzyl-amine, n-4-fluorophenyl methyl cyclopropanamine, cyclopropyl-4-fluorobenzyl amine, benzenemethanamine, n-cyclopropyl-4-fluoro, n-cyclopropyl-p-fluorobenzylamine, acmc-209n4r, n-cyclopropyl-4-fluorobenzylamine, n-4-fluorobenzyl cyclopropanamine #, n-4-fluorophenyl methyl cyclopropylamine
573031
N-[(4-fluorophenyl)methyl]cyclopropanamine
FC1=CC=C(CNC2CC2)C=C1
This item is not returnable. View return policy
625437-46-9
C10H12FN
165.21
MFCD07330007
YKNFXOCMGURJMK-UHFFFAOYSA-N
n-4-fluorobenzyl cyclopropanamine, cyclopropyl-4-fluoro-benzyl-amine, n-4-fluorophenyl methyl cyclopropanamine, cyclopropyl-4-fluorobenzyl amine, benzenemethanamine, n-cyclopropyl-4-fluoro, n-cyclopropyl-p-fluorobenzylamine, acmc-209n4r, n-cyclopropyl-4-fluorobenzylamine, n-4-fluorobenzyl cyclopropanamine #, n-4-fluorophenyl methyl cyclopropylamine
573031
N-[(4-fluorophenyl)methyl]cyclopropanamine
FC1=CC=C(CNC2CC2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2735
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material N-Cyclopropyl-4-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'