missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cyclopropyl-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60021.03

Code : NEW

Additional Details : CAS Number : 400839-43-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476488

  • £358.49 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Cyclopropyl-2-nitrobenzenesulfonamide
400839-43-2
244.25
MFCD00458271

Specifications

N-Cyclopropyl-2-nitrobenzenesulfonamide
Solid
244.25
1g
400839-43-2
97%
MFCD00458271
Videos
SDS